missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Carbonate Basic, Honeywell Fluka™

Product Code. 15611060 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg
Unit Size:
1kg
50kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611060 6 x 1 kg Pack of 6
15691050 1 kg 1kg
15601060 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15611060 Supplier Honeywell Research Chemicals Supplier No. 115136X1KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., white lead,≥77% Pb basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155

Specifications

Quantity 6 x 1 kg
Linear Formula (PbCO3)2·Pb(OH)2
UN Number UN3077
Formula Weight 775.63
Percent Purity ≥77% (Pb basis)
Grade puriss.
Chemical Name or Material Lead(Ii) Carbonate Basic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.