missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Carbonate Basic, Pure, SLR, Precipitated, Fisher Chemical™

Product Code. 10406880 Shop All Fisher Chemical Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
500 g
missing translation for 'unitSize':
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10406880 500 g 500g
10754951 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10406880 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' L/1150/53
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
MDL Number 78155

Specifications

Melting Point 400°C
Color White
Physical Form Powder/Solid
Quantity 500 g
Packaging Plastic powder jar
Density Vapor Density: 9.21
Formula Weight 775.59g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) carbonate, basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.