missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) iodide, 99.9985% (metals basis)

Product Code. 11387517
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387517 100 g 100g
11377517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11387517 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 012724.22
 Restricted Item
Estimated Shipment: 30-11-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 10101-63-0 | I2Pb | 461.00 g/mol

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Spécification

Boiling Point 954°C
Melting Point 402°C
Color Yellow
Physical Form Powder
Quantity 100 g
Odor Odorless
Sensitivity Light sensitive
Assay Percent Range 99.9985% (Metals basis)
Density 6.16
Solubility Information Soluble in concentrated solutions of alkali iodides and sodium thiosulfate. Insoluble in alcohol,cold hydrochloric acid and organic solvents.
Formula Weight 461
Chemical Name or Material Lead(II) iodide
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Lead(II) iodide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informations supplémentaires
  • MIXTURE LIST-Contain: Lead iodide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.