missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) iodide, ultra dry, 99.999% (metals basis)

Product Code. 11478410
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478410 100 g 100g
11458410 5 g 5g
11468410 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478410 Supplier Thermo Scientific Alfa Aesar Supplier No. 044314.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lead(II) Iodide, Ultra Dry, I2Pb, CAS Number-10101-63-0, blei ii-iodid, diiodo-, e2-plumbane, hsdb 636, lead diiodide, lead ii iodide, lead iodide pbi2, pbi2, plumbous iodide, plumbum jodatum, 100g, 99.999% (Metals basis), 954 deg.C, Yellow, 6.16, DOT Class: 6.1, Packaging Group: III, 233-256-9, 461 | CAS: 10101-63-0 | I2Pb | 461.00 g/mol

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Boiling Point 954°C
Melting Point 402°C
Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999% (Metals basis)
UN Number 2291
Solubility Information Soluble in concentrated solutions of alkali iodides and sodium thiosulfate. Insoluble in alcohol,cold hydrochloric acid and organic solvents.
Formula Weight 461
Odor Odorless
Sensitivity Light sensitive
Density 6.16
Chemical Name or Material Lead(II) Iodide, Ultra Dry
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Lead iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.