missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) metaborate monohydrate, Thermo Scientific Chemicals

Product Code. 11338108
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338108 100 g 100g
11328108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338108 Supplier Thermo Scientific Alfa Aesar Supplier No. 039339.22
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Lead(II) metaborate monohydrate acts as a starting material used in the preparation of lead(II) borates such as Pb2B3O5.5 (OH)2 and [Pb3(B3O7)](NO3) by changing the pH values of the reaction systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10214-39-8
MDL Number MFCD00049504
Synonym Lead(II) borate

Specifications

Melting Point 160°C (in -H2O)
Physical Form Powder
Quantity 100 g
Linear Formula PbB2O4·H2O
UN Number UN2291
Solubility Information Insoluble in water and alkali; Soluble in acids.
Formula Weight 310.81 (292.81 Anhydrous)
Odor Odorless
Density 5.598 g/mL
Chemical Name or Material Lead(II) metaborate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.