missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Fluka™

Product Code. 15914770 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity: 500 g; Packaging: Plastic Bottle; Grade: ACS Grade Salts
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15914770 Quantity: 500 g; Packaging: Plastic Bottle; Grade: ACS Grade Salts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15914770 Supplier Honeywell-Fluka Supplier No. 15338500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ≥99.0% (KT), yellow

  • Leading the industry for over 65 years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1.470°C (1.013 hPa)
Chemical Name or Material Lead(II) oxide
Melting Point 890°C
CAS 1317-36-8
Color Yellow
Quantity 500 g
Molecular Formula PbO
Grade ACS
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.