missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Oxide, ReagentPlus™, ≥99.9% trace metals basis, Honeywell™

Product Code. 15641940 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15641940 100 g Poly Bottle 100g
15651940 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15641940 Supplier Honeywell Research Chemicals Supplier No. 211907100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, <10 μm, Reagent Grade,≥99.9% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]

Specifications

Boiling Point 1470°C
Melting Point 890°C
Color Yellow
Physical Form Powder
Quantity 100 g
Linear Formula PbO
UN Number UN2291
Formula Weight 223.2g/mol
Percent Purity ≥99.9% (trace metals basis)
Grade ReagentPlus
Packaging Poly Bottle
Vapor Pressure 10mmHg (0°C)
Density 9.66g/cu.cm
Chemical Name or Material Lead(Ii) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.