missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Sulfide, Pure, Fisher Chemical

Product Code. 10264150 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10264150 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10264150 Supplier Fisher Chemical Supplier No. L/1730/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100g, Glass Jar

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1281.1°C
Melting Point 1113.9°C
CAS 1314-87-0
Color Silver
Physical Form Solid
Quantity 100 g
Molecular Formula PbS
MDL Number MFCD00016280
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES [S--].[Pb++]
IUPAC Name λ²-lead(2+) sulfanediide
Molecular Weight (g/mol) 239.30
PubChem CID 14819
Formula Weight 239.26g/mol
Packaging Glass Jar
Vapor Pressure &1mmHg&852
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.