missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) fluoride, 99%

Product Code. 15441037
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441037 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441037 Supplier Thermo Scientific Alfa Aesar Supplier No. 013070.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-59-7 | F4Pb | 283.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-59-7
Physical Form Crystalline
Quantity 2 g
Molecular Formula F4Pb
MDL Number MFCD00016278
UN Number UN2291
Synonym lead iv fluoride, lead tetrafluoride, pbf4, tetrafluoridolead, blei iv-fluorid
InChI Key YAFKGUAJYKXPDI-UHFFFAOYSA-J
SMILES F[Pb](F)(F)F
IUPAC Name tetrafluoroplumbane
Molecular Weight (g/mol) 283.194
PubChem CID 123258
ChEBI CHEBI:37186
Formula Weight 283.18
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Lead(IV) fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(IV) fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.