missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) Oxide, ≥97.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15652360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15652360 100 g Poly Bottle 100g
15662360 500 g Poly Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15652360

Brand: Honeywell-Fluka 237140100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥97.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O

Specifications

Color Black
Quantity 100 g
Linear Formula PbO2
UN Number UN1872
Formula Weight 239.2g/mol
Percent Purity ≥97.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Lead(Iv) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.