missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97%

Product Code. 11460703
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
Product Code. Quantity unitSize
11460703 500 g 500g
11470703 2500 g 2500g
2 options
This item is not returnable. View return policy

Product Code. 11460703

Brand: Thermo Scientific Alfa Aesar A12742.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.

Solubility
Insoluble in water. Soluble in dil.HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Quantity 500 g
UN Number UN1872
Merck Index 14,5407
Solubility Information Insoluble in water. Soluble in dil.HCl.
Formula Weight 239.19
Percent Purity 97%
Odor Odorless
Density 9.38
Chemical Name or Material Lead(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.