missing translation for 'onlineSavingsMsg'
Learn More

Lead Nitrate, 0.1M, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11414799 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1000 mL
Packaging:
Tamper evident
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11414799 1000 mL Tamper evident 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414799 Supplier Reagecon Supplier No. 3003100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.2.2) and prepared in an ISO 9001 facility. Lead Nitrate, 0.1M, EP Grade, Reagecon™ is a ready-to-use volumetric solution with 33g/L Pb(NO3)2 in water for use as a laboratory reagent and oxidizing agent.

Lead nitrate is a water-soluble lead(II) salt. It is a toxic compound that functions as an oxidizing agent and is used in making other lead compounds. It disassociates into its constituent ions, Pb2+ (aq) and NO3.  This solution is prepared by dissolving 33g of lead nitrate into water and diluting to 1L.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.2.2) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise volumetric solutions

Specifications

CAS 10099-74-8, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 1000 mL
Molecular Formula N2O6Pb
MDL Number MFCD00011153
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
Solubility Information Soluble in water
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name lead(2+);dinitrate
Molecular Weight (g/mol) 331.20
PubChem CID 24924
ChEBI CHEBI:37187
Formula Weight 331.21g/mol
Grade Eu. Ph. Chapter 4(4.2.2)
Concentration or Composition (by Analyte or Components) 33g lead nitrate per 1L
Odor Odorless
Packaging Tamper evident
Chemical Name or Material Lead(II) nitrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 0.1M LEAD NITRATE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH201-Contains lead. Should not be used on surfaces liable to be chewed or sucked by children.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : lead dinitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.