missing translation for 'onlineSavingsMsg'
Learn More

Lead Subacetate, ACS Reagent, Honeywell™ Fluka™

Product Code. 15672360 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15672360

Brand: Honeywell-Fluka 237159500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1335-32-6
Physical Form Solid
Quantity 500 g
Linear Formula Pb(C2H3O2)2 · 2Pb[OH]2
MDL Number MFCD00013041
UN Number UN1616
Formula Weight 807.72g/mol
Grade ACS Reagent
Packaging Poly bottle
Chemical Name or Material Lead Subacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.