missing translation for 'onlineSavingsMsg'
Learn More

Lead Tetraacetate, purum p.a., ≥95% (dried material), Honeywell Burdick & Jackson™

Product Code. 15671450 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671450 500 g Glass Bottle 500g
15661450 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15671450 Supplier Honeywell Research Chemicals Supplier No. 15370500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum p.a., moistened with ∼5% acetic acid to stabilize,≥95% (dried material)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-67-8
Molecular Formula C8H16O8Pb
Molecular Weight (g/mol) 447.408
MDL Number MFCD00008693
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Melting Point 180° to 190°
Color White
Quantity 500 g
Linear Formula Pb(CH3COO)4
UN Number UN2923
Beilstein 3595640
Formula Weight 443.38g/mol
Percent Purity ≥95% (dried material)
Grade purum p.a.
Packaging Glass Bottle
Appearance Crystals
Density 2.230g/cu.cm
Chemical Name or Material Lead Tetraacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.