missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Leaf Weight

Product Code. 16383136 Shop All Sartorius Products
Change view
Click to view available options
Weight (Metric):
1 mg
5 mg
20 mg
50 mg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
16383136 1 mg Each
16303146 5 mg Each
16313146 20 mg Each
16323146 50 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16383136 Supplier Sartorius Supplier No. YCW01302
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Leaf Weight is available in aluminum or nickel silver and comes in a case for protection.

  • F1 weights sets from 1 mg to 10 kg
  • Up to 500 mg leaf weights, Material: Aluminium, Density: 2.7 g/cm3
  • Leaf weights 1 to 5 g, Material: Nickel Silver, Density: 8.7 g/cm3
  • Leaf weights 10 to 500 mg, Material: Special steel, non-magnetizable, density 8.0 g/cm3, highly corrosion-resistant, knob weights highly polished, in wooden case
  • Packaging of the weights:
    • From 1 mg up to 500 mg in a plastic case
    • Up to 20 g in a plastic case
    • From 50 g and up in a wooden case
    • From 1 kg, additionally supplied with gloves
    • Service weight set in a plastic carrying case
TRUSTED_SUSTAINABILITY

Specifications

Product Type Calibration Mass
Certifications/Compliance DAkkS Certificate
Classification OIML F1
Material Aluminum, Nickel Silver, Stainless Steel
Weight (Metric) 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.