missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MedChemExpress

Product Code. 16492306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492306 500 mg 500mg
16468876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492306 Supplier Medchem Express Supplier No. HYB2235
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.

Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.

Specifications

Chemical Name or Material Lecithin
CAS 8002-43-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C42H80NO8P
Quantity 500 mg
Solubility Information DMSO : 5 mg/mL (6.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (4.39 mM; Need ultrasonic)
SMILES O=P(OC[C@@H](COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)(OCC[N+](C)(C)C)[O-]
Molecular Weight (g/mol) 758.06
Formula Weight 758.06
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.