Learn More
Berthold Technologies LED Plant Growth Illumination Far-red/R/G/B
Product Code. 31777393 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
LED Panel
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777393 LED Panel Each
1 options
Product Code. 31777393 Supplier Berthold Technologies Supplier No. 5658901
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
LED illumination for NightSHADE imaging systems that enables daylight simulation with customizable settings.
  • Two LED panels, each with four colors
  • Individual control of LED intensity and duration
  • Supports simulation of daylight with spectral and intensity gradients
  • Equipped with the following colors: Far red, Red, Green, Blue
Specifications
Type LED Panel
For Use With (Application) Daylight Simulation
For Use With (Equipment) LED Panel
Voltage 24 V