missing translation for 'onlineSavingsMsg'
Learn More

Ledipasvir (acetone), MedChemExpress

Product Code. 16489227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489227

Brand: Medchem Express HY15602A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ledipasvir acetone (GS-5885 acetone) is the active ingredient of Ledipasvir. Ledipasvir is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon.

Ledipasvir acetone (GS-5885 acetone) is the active ingredient of Ledipasvir. Ledipasvir is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon.

Specifications

Chemical Name or Material Ledipasvir (acetone)
CAS 1441674-54-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C52H60F2N8O7
Quantity 5 mg
Synonym GS-5885 acetone
Solubility Information DMSO : 110 mg/mL (116.15 mM; Need ultrasonic)
SMILES O=C(OC)N[C@H](C(N([C@H](C1=NC=C(C2=CC(C(F)(F)C3=C4C=CC(C5=CC=C6N=C([C@H]7N(C([C@@H](NC(OC)=O)C(C)C)=O)[C@]8([H])CC[C@@]7([H])C8)NC6=C5)=C3)=C4C=C2)N1)C9)CC%109CC%10)=O)C(C)C.CC(C)=O
Molecular Weight (g/mol) 947.08
Formula Weight 947.08
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.