missing translation for 'onlineSavingsMsg'
Learn More

LEE 011-d6, TRC

Product Code. 30477513
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477513 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477513 Supplier Toronto Research Chemicals Supplier No. TRCL32100210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-cyclopentyl-2-[(5-piperazin-1-ylpyridin-2-yl)amino]-N,N-bis(trideuteriomethyl)pyrrolo[2,3-d]pyrimidine-6-carboxamide
CAS 1328934-40-2
Molecular Formula C23 D6 H24 N8 O
Quantity 10 mg
Synonym LEE 011-d6, 7-Cyclopentyl-N,N-di(methyl-d3)-2-[[5-(1-piperazinyl)-2-pyridinyl]amino]-7H-Pyrrolo[2,3-d]pyrimidine-6-carboxamide, Ribociclib-d6
InChI Formula InChI=1S/C23H30N8O/c1-29(2)22(32)19-13-16-14-26-23(28-21(16)31(19)17-5-3-4-6-17)27-20-8-7-18(15-25-20)30-11-9-24-10-12-30/h7-8,13-15,17,24H,3-6,9-12H2,1-2H3,(H,25,26,27,28)/i1D3,2D3
SMILES [2H]C([2H])([2H])N(C(=O)c1cc2cnc(Nc3ccc(cn3)N4CCNCC4)nc2n1C5CCCC5)C([2H])([2H])[2H]
IUPAC Name 7-cyclopentyl-2-[(5-piperazin-1-ylpyridin-2-yl)amino]-N,N-bis(trideuteriomethyl)pyrrolo[2,3-d]pyrimidine-6-carboxamide
Molecular Weight (g/mol) 440.574
Formula Weight 440.292
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.