missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Legionella 1 Latex Reagent

Product Code. 17303201 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17303201 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17303201 Supplier Novacyt - Microgen Bioproducts Supplier No. M45A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microgen Bioproducts Legionella 1 Latex reagent

  • Individual L.pneumophila serogroups 1, 2-15, and non L.pneumophila species latex and positive control available separately.

Spezifikation

Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes L. pneumophila Serogroup 1 Antibodies
Format 1 Bottle (2.5 mL)
Sample Type Bacterial Colonies
Sensitivity 1
Type Reagent
For Use With (Application) The Confirmatory Identification of Legionella Pneumophila and Other Commonly Isolated Legionellasp Ecies Grown on Selective Medi
For Use With (Equipment) 0.85% Isotonic Saline, Disposable Agglutination Slides, Disposable Mixing Sticks, Bacteriological Loops, Legionella Selective Medium, Glass Tubes for Boiling
Includes 2.5 mL
Material Polyethylene Dropper Bottle
No. of Reactions 1/Test
Organism Group Monera
Packaging Type Bottle in polygrip bag
Quantity 2.5 mL
Reagent Type M45a Legionella Test Reagent 1 x 2.5 mL
Species Legionella pneumophila
Specificity 1
Target Organism Class Bacteria
Test Time 7 minutes
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.