missing translation for 'onlineSavingsMsg'
Learn More

Brannan Legionnaire Kit In A Carry Case

Product Code. 32179304
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32179304 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32179304 Supplier Brannan Supplier No. 13/800/3
  • ONLINE EXCLUSIVE PRICE
    198.14 €
    107.18 € / Pack of 1
    Save 90.96 €
    45% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for checking water in pipes and tanks forming part of a water system.

  • Includes fully waterproof thermometer with a factory calibrated certificate
  • Clip on Pipe thermometer 0°C to 120°C
  • Water resistant timer IP64, with suction cup fixture
  • Disposable gloves
  • Antiseptic wipes (10 Pc.)
  • Operating instructions
  • All products supplied in a carry case

Specifications

CE Marker Yes
Detectable Analytes Legionella
Type Legionella Test Kit
For Use With (Application) Test Hot and Cold Water Systems for Legionella
Includes Waterproof Thermometer with Factory Calibration Certificate, Clip-On Pipe Thermometer, Digital Water-Resistant Timer, Disposable Gloves, Antiseptic Wipes
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.