missing translation for 'onlineSavingsMsg'
Learn More

Lemborexant, TRC

Product Code. 30465656
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465656 100 mg 100mg
30469936 10 mg 10mg
30482083 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465656 Supplier Toronto Research Chemicals Supplier No. TRCL215945100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1369764-02-2
Molecular Formula C22H20F2N4O2
Molecular Weight (g/mol) 410.42
Synonym (1R,2S)-2-[[(2,4-Dimethyl-5-pyrimidinyl)oxy]methyl]-2-(3-fluorophenyl)-N-(5-fluoro-2-pyridinyl)cyclopropanecarboxamide
IUPAC Name (1R,2S)-2-[(2,4-dimethylpyrimidin-5-yl)oxymethyl]-2-(3-fluorophenyl)-N-(5-fluoropyridin-2-yl)cyclopropane-1-carboxamide
SMILES Cc1ncc(OC[C@]2(C[C@H]2C(=O)Nc3ccc(F)cn3)c4cccc(F)c4)c(C)n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H20F2N4O2/c1-13-19(11-25-14(2)27-13)30-12-22(15-4-3-5-16(23)8-15)9-18(22)21(29)28-20-7-6-17(24)10-26-20/h3-8,10-11,18H,9,12H2,1-2H3,(H,26,28,29)/t18-,22+/m0/s1
Formula Weight 410.1554
Percent Purity >95%
Chemical Name or Material Lemborexant
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.