missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lenalidomide 5'-piperazine-4-methylpiperidine
GREENER_CHOICE

Product Code. 18745806
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18745806 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18745806 Supplier Tocris Bioscience™ Supplier No. 7473/25
  • ONLINE EXCLUSIVE PRICE
    375.00 €
    353.85 € / 25mg
    Save 21.15 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cereblon ligand with terminal piperidine for onward chemistry

Lenalidomide 5'-piperazine-4-methylpiperidine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand with a terminal piperidine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[1,3-Dihydro-1-oxo-5-[4-(4-piperidinylmethyl)-1-piperazinyl]-2 H-isoindol-2-yl]-2,6-piperidinedione dihydrochloride
CAS 2520105-60-4
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C23H31N5O30.2 HCl
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.