missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lenalidomide, 98%

Product Code. 16544344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16544344 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16544344 Supplier Thermo Scientific™ Supplier No. 461590010
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 191732-72-6 | C13H13N3O3 | 259.27 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 191732-72-6
Assay Percent Range 98%
Molecular Formula C13H13N3O3
Quantity 1 g
InChI Key GOTYRUGSSMKFNF-UHFFFAOYNA-N
SMILES NC1=CC=CC2=C1CN(C1CCC(=O)NC1=O)C2=O
IUPAC Name 3-(4-amino-1-oxo-2,3-dihydro-1H-isoindol-2-yl)piperidine-2,6-dione
Molecular Weight (g/mol) 259.27
Formula Weight 259.3
Chemical Name or Material Lenalidomide
compliance-icons
Product Identifier
  • Lenalidomide, 98%
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lenalidomide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.