missing translation for 'onlineSavingsMsg'
Learn More

Lenalidomide, MedChemExpress

Product Code. 16457976
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457976 100 mg 100mg
16441999 1 g 1g
16441966 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457976 Supplier Medchem Express Supplier No. HYA0003
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lenalidomide (CC-5013), a derivative of Thalidomide, acts as molecular glue.

Lenalidomide is an orally active immunomodulator. Lenalidomide (CC-5013) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide (CC-5013) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells.

Specifications

Chemical Name or Material Lenalidomide
CAS 191732-72-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H13N3O3
Quantity 100 mg
Synonym CC-5013
Solubility Information DMSO : 100 mg/mL (385.71 mM; Need ultrasonic)
SMILES O=C1C2=CC=CC(N)=C2CN1C(C(N3)=O)CCC3=O
Molecular Weight (g/mol) 259.26
Formula Weight 259.26
Percent Purity 99.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.