missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lenalidomide

Product Code. 30000855
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000855 Supplier Tocris Bioscience™ Supplier No. 6305/100
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cereblon binder; induces ubiquitination and degradation of CK1α by E3 ubiquitin ligase

Thalidomide analog. Immune modulatory drug and cereblon binding compound. Induces ubiquitination and degradation of casein kinase (CK) 1α by the E3 ubiquitin ligase CRL4CRBN. Also TNF-α inhibitor and angiogenesis inhibitor. Promotes degradation of transcription factor SALL4.
TRUSTED_SUSTAINABILITY

Specifications

CAS 191732-72-6
Quantity 100 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C13H13N3O3
Formula Weight 259.26
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 259.265
Chemical Name or Material 3-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.