missing translation for 'onlineSavingsMsg'
Learn More

Lenalidomide, TRC

Product Code. 18091410
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091410 500 mg 500mg
18041520 100 mg 100mg
17989939 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091410 Supplier Toronto Research Chemicals Supplier No. TRCL328000500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191732-72-6
Molecular Formula C13 H13 N3 O3
Molecular Weight (g/mol) 259.26
Synonym 3-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione, 1-Oxo-4-amino-2-(2,6-dioxopiperidin-3-yl)isoindole, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidin-2,6-dione, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione, CC 5013, CDC 501, ENMD 0997, Lenalidomide, Revimid, Revlimid
IUPAC Name 3-(7-amino-3-oxo-1H-isoindol-2-yl)piperidine-2,6-dione
SMILES Nc1cccc2C(=O)N(Cc12)C3CCC(=O)NC3=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H13N3O3/c14-9-3-1-2-7-8(9)6-16(13(7)19)10-4-5-11(17)15-12(10)18/h1-3,10H,4-6,14H2,(H,15,17,18)
Formula Weight 259.0957
Percent Purity >95%
Chemical Name or Material Lenalidomide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.