missing translation for 'onlineSavingsMsg'
Learn More

Lenperone, TRC

Product Code. 30478179
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478179 50 mg 50mg
30474497 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478179 Supplier Toronto Research Chemicals Supplier No. TRCL32880050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24678-13-5
Molecular Formula C22 H23 F2 N O2
Molecular Weight (g/mol) 371.42
Synonym 4-[4-(4-Fluorobenzoyl)-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone
IUPAC Name 4-[4-(4-fluorobenzoyl)piperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
SMILES Fc1ccc(cc1)C(=O)CCCN2CCC(CC2)C(=O)c3ccc(F)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H23F2NO2/c23-19-7-3-16(4-8-19)21(26)2-1-13-25-14-11-18(12-15-25)22(27)17-5-9-20(24)10-6-17/h3-10,18H,1-2,11-15H2
Formula Weight 371.1697
Chemical Name or Material Lenperone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.