Learn More
Invitrogen™ LentiArray™ Cas9 Lentivirus
Description
LentiArray Cas9 Lentivirus provides an efficient method to drive high level Cas9 nuclease expression in a wide variety of cell types. The LentiArray Cas9 Lentivirus construct includes a human codon-optimized version of Cas9 with two nuclear localization signals to facilitate efficient delivery into the nucleus. The 100 μL size is sufficient for creating stable Cas9 expressing pools of cells or for isolating stable Cas9 expressing clones. LentiArray Cas9 Lentivirus is provided as ready to use lentivirus at a titer of 1 x 107 TU/mL.
Recommended Storage
Store at -68 to -85°C
Specifications
Specifications
| Content And Storage | 1 x 100 μL. Store at -68 to -85°C. |
| Delivery Type | Lentiviral |
| Sample Type | gRNA |
| Promoter | EF-1a |
| Transfection Technique | Lentiviral transduction |
| Species | Human |
| For Use With (Application) | CRISPR gene knockout |
| Product Line | LentiArray |
| Product Type | Cas9 Lentivirus |
| Quantity | 100 μL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.