missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP

Product Code. 15697888
Change view
Click to view available options
Quantity:
100 μL
1 mL
Unit Size:
100µL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15697888 100 μL 100µL
15910383 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15697888 Supplier Invitrogen™ Supplier No. A32060
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%.

The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

Recommended Storage

Store at -68 to -85°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 100 μL. Store (at -68°C to -85°C.)
Delivery Type Lentiviral
Format Tube
Control Type Positive Control
RNAi Type gRNA
Species Human
For Use With (Application) Genome Editing
Product Line LentiArray
Product Type gRNA
Promoter U6
Quantity 100 μL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.