Learn More
Emma Safety Footwear Leo XD Safety Shoes
Product Code. 15998851
Change view
Click to view available options
Size:
37
38
39
47
48
49
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998851 39 1 pair
15978851 37 1 pair
15988851 38 1 pair
15908861 47 1 pair
15918861 48 1 pair
15928861 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15998851 Supplier Emma Safety Footwear Supplier No. MM70156839
  • ONLINE EXCLUSIVE PRICE
    122.85 €
    77.02 € / 1 pairs
    Save 45.83 €
    37% Off
View all promotions
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip.  Available in a variety of widths.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Leo XD
Material Leather
Size 39