missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Leptin Protein, Human
Specifications
Specifications
| Content And Storage | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Quantity | 100 μg |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | Approximately 16.0 kDa |
| Synonym | rHuLeptin∣ Obesity protein∣ OB |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?