missing translation for 'onlineSavingsMsg'
Learn More

Leu-Enkephalin Amide H-Tyr-Gly-Gly-Phe-Leu-NH2 Hydrochloride Salt, TRC

Product Code. 30468395
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468395 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468395 Supplier Toronto Research Chemicals Supplier No. TRCL24716050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Leu-Enkephalin Amide H-Tyr-Gly-Gly-Phe-Leu-NH2 Hydrochloride Salt
CAS 99236-50-7
Molecular Formula C28H38N6O6 . HCl
Quantity 50 mg
Synonym L-Leucinamide, L-tyrosylglycylglycyl-L-phenylalanyl-, monohydrochloride (9CI), 5-L-Leucinamide-1-5-β-neoendorphin (human) hydrochloride, L-Tyrosylglycylglycyl-L-phenylalanyl-L-leucinamide monohydrochloride, Leucine enkephalin amide hydrochloride, Leu-enkephalin amide hydrochloride
InChI Formula InChI=1S/C28H38N6O6.ClH/c1-17(2)12-22(26(30)38)34-28(40)23(14-18-6-4-3-5-7-18)33-25(37)16-31-24(36)15-32-27(39)21(29)13-19-8-10-20(35)11-9-19;/h3-11,17,21-23,35H,12-16,29H2,1-2H3,(H2,30,38)(H,31,36)(H,32,39)(H,33,37)(H,34,40);1H/t21-,22-,23-;/m0./s1
SMILES Cl.CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N
IUPAC Name (2S)-2-[[(2S)-2-[[2-[[2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]acetyl]amino]acetyl]amino]-3-phenylpropanoyl]amino]-4-methylpentanamide;hydrochloride
Molecular Weight (g/mol) 591.1
Formula Weight 590.262
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.