missing translation for 'onlineSavingsMsg'
Learn More

Leucic Acid, TRC

Product Code. 30005640
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005640 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30005640 Supplier Toronto Research Chemicals Supplier No. TRCL3300801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 498-36-2
Molecular Formula C6H12O3
Quantity 1 g
Synonym (±)-2-Hydroxyisocaproic acid, (±)-2-Hydroxyisohexanoic acid, (±)-Leucic acid, (±)-Leucinic acid, (±)-α-Hydroxyisocaproic acid, 2-Hydroxy-4-methylpentanoic acid, 2-Hydroxy-4-methylvaleric acid, 2-Hydroxyisocaproic acid, 2-Hydroxyisohexanoic acid, DL-2-Hydroxy-4-methylpentanoic acid, DL-2-Hydroxyisocaproic acid, DL-Leucic acid, DL-α-Hydroxyisocaproic acid, Leucinic acid, α-Hydroxy-iso-caproic acid, α-Hydroxyisocaproate, α-Hydroxyisocaproic acid
InChI Formula InChI=1S/C6H12O3/c1-4(2)3-5(7)6(8)9/h4-5,7H,3H2,1-2H3,(H,8,9)
SMILES CC(C)CC(O)C(=O)O
Molecular Weight (g/mol) 132.16
Formula Weight 132.0786
Chemical Name or Material Leucic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.