missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leuco Crystal Violet, 99%, pure

Product Code. 10138742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10138742 5 g Glass bottle 5g
10781091 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10138742 Supplier Thermo Scientific Chemicals Supplier No. 204330050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 603-48-5 | C25H31N3 | 373.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym 4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
PubChem CID 69048
IUPAC Name 4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 175.0°C to 180.0°C
Assay Percent Range 99%
Linear Formula CH[C6H4N(CH3)2]3
Beilstein 13, 315
Infrared Spectrum Authentic
Formula Weight 373.53
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Color Gray to White
Quantity 5 g
Physical Form Powder
Chemical Name or Material Leuco Crystal Violet, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Leuco Crystal Violet
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.