missing translation for 'onlineSavingsMsg'
Learn More

Leucocrystal violet, 99.8%, For HPLC analysis, MP Biomedicals™

Product Code. 11473392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473392 25 g 25g
11463392 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473392 Supplier MP Biomedicals Supplier No. 0215519525
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym 4,4',4''-Methylidynetris(N,N-dimethylaniline)
PubChem CID 69048
IUPAC Name 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Color Gray/Purple
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 373.5
Percent Purity 99.8%
Grade Spectrophotometry
Chemical Name or Material Leucocrystal violet, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.