missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green

Product Code. 11380378
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11380378 2 g 2g
11370378 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380378 Supplier Thermo Scientific Chemicals Supplier No. 043758.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4,4'-Benzylidenebis(N,N-dimethylaniline); Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Melting Point 100°C to 102°C
Sensitivity Air Sensitive
Beilstein 2140506
Merck Index 14,5699
Solubility Information Soluble in water (<0.1mg/ml),chloroform,benzene,ethyl ether,ethanol and toluene.
Quantity 2 g
Physical Form Powder
Formula Weight 330.47
Chemical Name or Material Leucomalachite Green

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.