missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leucomalachite Green, pure

Product Code. 10781242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10781242 25 g Glass bottle 25g
10696801 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781242 Supplier Thermo Scientific Chemicals Supplier No. 402110250
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 129-73-7 | C23H26N2 | 330.475 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23H26N2
Molecular Weight (g/mol) 330.475
MDL Number MFCD00008315
InChI Key WZKXBGJNNCGHIC-UHFFFAOYSA-N
Synonym 4, 4';-Benzylidenebis(N, N-dimethylaniline), Malachite green leuco base
PubChem CID 67215
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=C(C=C3)N(C)C

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Beilstein 13, 275
Merck Index 15, 5763
Color Blue or Brown/Green to White
Quantity 25 g
Physical Form Powder
Formula Weight 330.46
Grade Pure
Chemical Name or Material Leucomalachite Green
Show More Show Less
compliance-icons
Product Identifier
  • Leucomalachite Green
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.