missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Leucyl-phenylalanine

Product Code. 16401047
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401047 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401047 Supplier MedChemExpress Supplier No. HY113278
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Leucyl-phenylalanine belongs to the class of organic compounds known as dipeptides.

Specifications

Purity 97.0%
CAS 3063-05-6
Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 278.35 g/mol
Product Type Natural Products
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : 4 mg/mL (14.37 mM∣Need ultrasonic) ∣Ethanol : 1.11 mg/mL (3.99 mM∣Need ultrasonic) ∣DMSO : <1 mg/mL (ultrasonic) (insoluble or slightly soluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.