missing translation for 'onlineSavingsMsg'
Learn More

Leucyl-Phenylalanyl-Isoleucyl-Glutamyl-Tryptophyl-Leucyl-Lysine xTFA Salt, TRC

Product Code. 30471672
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471672 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471672 Supplier Toronto Research Chemicals Supplier No. TRCL3301905MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Leucyl-Phenylalanyl-Isoleucyl-Glutamyl-Tryptophyl-Leucyl-Lysine xTFA Salt
CAS 868249-04-1
Molecular Formula C49H73N9O10• x(C2HF3O2)
Quantity 5 mg
Synonym Leu-Phe-Ile-Glu-Trp-Leu-Lys xTFA, LFIEWLK xTFA
InChI Formula InChI=1S/C7H14N2/c1-3-8-6-5-7-9-4-2/h3-4,8-9H,1-2,5-7H2
SMILES C=CNCCCNC=C
IUPAC Name N,N'-bis(ethenyl)propane-1,3-diamine
Molecular Weight (g/mol) 948.1611402
Formula Weight 126.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.