missing translation for 'onlineSavingsMsg'
Learn More

Leucylisoleucine, TRC

Product Code. 30179246
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179246 500 mg 500mg
30179183 100 mg 100mg
30178958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179246 Supplier Toronto Research Chemicals Supplier No. TRCA899030500MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36077-41-5
Synonym (2S,3S)-2-((S)-2-Amino-4-methylpentanamido)-3-methylpentanoic Acid∣L-Leucyl-L-isoleucine∣165: PN: WO2011146121 PAGE: 117 Claimed Sequence

Specifications

Format Neat
Molecular Weight (g/mol) 244.33
Quantity 500 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.