missing translation for 'onlineSavingsMsg'
Learn More

Leupeptin Trifluoroacetate Salt, MP Biomedicals

Product Code. 11442491 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442491 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442491 Supplier MP Biomedicals Supplier No. 0219562325
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 147385-61-3
Molecular Formula C22H39F3N6O6
Molecular Weight (g/mol) 540.59
MDL Number MFCD00133458
InChI Key WUHGBZVQELGOMH-UHFFFAOYNA-N
Synonym leupeptin trifluoroacetate salt, n-5-carbamimidamido-1-oxopentan-2-yl-2-2-acetamido-4-methylpentanamido-4-methylpentanamide; trifluoroacetic acid, trifluoroacetic acid-n-acetylleucyl-n-5-diaminomethylidene amino-1-oxopentan-2-yl leucinamide 1/1
PubChem CID 44134799
IUPAC Name N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide; trifluoroacetic acid
SMILES OC(=O)C(F)(F)F.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O

Specifications

Quantity 25 mg
Formula Weight 540.6
Physical Form Powder
Chemical Name or Material Leupeptin Trifluoroacetate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.