missing translation for 'onlineSavingsMsg'
Learn More

Levamisole hydrochloride, 99.62%, MP Biomedicals™

Product Code. 11493412 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493412 5 g 5g
11403422 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493412 Supplier MP Biomedicals Supplier No. 0215522805
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16595-80-5
Molecular Formula C11H13ClN2S
Molecular Weight (g/mol) 240.749
InChI Key LAZPBGZRMVRFKY-HNCPQSOCSA-N
Synonym levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
PubChem CID 27944
ChEBI CHEBI:6433
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
SMILES C1CSC2=NC(CN21)C3=CC=CC=C3.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 240.8
Percent Purity 0.9962
Physical Form Crystalline Powder
Chemical Name or Material Levamisole hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.