missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Levetiracetam

Product Code. 18742594
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18742594 50 mg 50mg
18702494 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18742594 Supplier R&D Systems™ Supplier No. 2839/50
  • ONLINE EXCLUSIVE PRICE
    588.29 €
    530.25 € / 50mg
    Save 58.04 €
    9% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antiepileptic; binds SV2A

Levetiracetam is an antiepileptic that displays distinctive properties from conventional antiepileptic drugs. Displays potent seizure protection in animal models of chronic epilepsy but lacks activity in acute seizure models. Binds synaptic vesicle protein 2A (SV2A) and inhibits Na+-dependent Cl-/HCO3- exchange. In a mouse model of Alzheimer's disease, Levetiracetam rescues levels of presynaptic vesicle fusion and neurotransmitter release, reduces Aβ42 and APP levels and decreases deposits of amyloid plaque; in vitro Levetiracetam inhibits Aβ-induced glutamate from astrocytes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 170.21
Purity >99%
Molecular Weight (g/mol) 170.21
Chemical Name or Material Levetiracetam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.