missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam

Product Code. 15701799 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15701799

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102767-28-2 | C8H14N2O2 | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD03265610
InChI Key HPHUVLMMVZITSG-ZCFIWIBFSA-N
Synonym levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
PubChem CID 5284583
ChEBI CHEBI:6437
IUPAC Name (2R)-2-(2-oxopyrrolidin-1-yl)butanamide
SMILES CC[C@@H](N1CCCC1=O)C(N)=O

Specifications

Melting Point 117.0°C
Quantity 5 g
Specific Rotation -76
Solubility Information Solubility in water: 104 g/100 mL. Other solubilities: soluble in chloroform, methanol, acetone
Formula Weight 170.21
Chemical Name or Material Levetiracetam
compliance-icons
Product Identifier
  • Levetiracetam
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Levetiracetam

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.