missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam

Product Code. 15711799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15711799 25 g 25g
15701799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15711799 Supplier Thermo Scientific Acros Supplier No. 462120250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102767-28-2 | C8H14N2O2 | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD03265610
InChI Key HPHUVLMMVZITSG-ZCFIWIBFSA-N
Synonym levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
PubChem CID 5284583
ChEBI CHEBI:6437
IUPAC Name (2R)-2-(2-oxopyrrolidin-1-yl)butanamide
SMILES CC[C@@H](N1CCCC1=O)C(N)=O

Specifications

Melting Point 117.0°C
Quantity 25 g
Specific Rotation -76
Solubility Information Solubility in water: 104 g/100 mL. Other solubilities: soluble in chloroform, methanol, acetone
Formula Weight 170.21
Chemical Name or Material Levetiracetam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.