missing translation for 'onlineSavingsMsg'
Learn More

Levulinic acid, 98%

Product Code. 11429752
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429752 250 g 250g
11419752 1000 g 1000g
11439752 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429752 Supplier Thermo Scientific Alfa Aesar Supplier No. A10813.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-76-2 | C5H8O3 | 116.116 g/mol

Mainly used as an intermediate for medicines, agricultural chemical and organic synthesis. Used as raw material of perfume, modifier and solvent of plastics. It is also employed as an additive of polymer, paints and lubricating oil. Also used as a surfactant and auxiliary for making printing ink, rubber flux and cosmetics(including shampoo).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used as an intermediate for medicines, agricultural chemical and organic synthesis. Used as raw material of perfume, modifier and solvent of plastics. It is also employed as an additive of polymer, paints and lubricating oil. Also used as a surfactant and auxiliary for making printing ink, rubber flux and cosmetics(including shampoo).

Solubility
Soluble in water ( 675g/L at 20°C).

Notes
Light Sensitive. Store in dark. Store away from oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-76-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00002796
InChI Key JOOXCMJARBKPKM-UHFFFAOYSA-N
Synonym levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid
PubChem CID 11579
ChEBI CHEBI:45630
IUPAC Name 4-oxopentanoic acid
SMILES CC(=O)CCC(=O)O

Specifications

Melting Point 26°C to 31°C
Density 1.134
pH 2.5 to 3.0
Boiling Point 245°C to 246°C
Flash Point 137°C (278°F)
Odor Characteristic
Refractive Index 1.4415
Quantity 250 g
UN Number UN3261
Beilstein 506796
Sensitivity Light sensitive
Merck Index 14,5472
Solubility Information Soluble in water ( 675g/L at 20°C).
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material Levulinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.