missing translation for 'onlineSavingsMsg'
Learn More

Lexibulin, MedChemExpress

Product Code. 16490357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490357 50 mg 50mg
16466717 5 mg 5mg
16444008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490357 Supplier Medchem Express Supplier No. HY10498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lexibulin (CYT-997) is a potent and orally active tubulin polymerisation inhibitor with IC50s of 10-100 nM in cancer cell lines; with potent cytotoxic and vascular disrupting activity in vitro and in vivo. Lexibulin induces cell apoptosis and induces mitochondrial ROS generation in GC cells.

Lexibulin (CYT-997) is a potent and orally active tubulin polymerisation inhibitor with IC50s of 10-100 nM in cancer cell lines

Specifications

Chemical Name or Material Lexibulin
CAS 917111-44-5
Color White
Purity Grade Notes Research
Molecular Formula C24H30N6O2
Quantity 50 mg
Synonym CYT-997
Solubility Information DMSO : ≥ 100 mg/mL (230.13 mM)
SMILES CCC[C@H](NC1=C(C)C=NC(C2=CC=C(NC(NCC)=O)C(OC)=C2)=N1)C3=CN=CC=C3
Molecular Weight (g/mol) 434.53
Formula Weight 434.53
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.