missing translation for 'onlineSavingsMsg'
Learn More

Lexithromycin, TRC

Product Code. 30473425
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473425 25 mg 25mg
30466335 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473425 Supplier Toronto Research Chemicals Supplier No. TRCL39770325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lexithromycin
CAS 53066-26-5
Molecular Formula C38 H70 N2 O13
Quantity 25 mg
Synonym Erythromycin A 9-(O-methyloxime), Erythromycin A 9-Methoxime, Wy 48314, Oxacyclotetradecane, Erythromycin deriv.
InChI Formula InChI=1S/C38H70N2O13/c1-15-26-38(10,46)31(42)21(4)28(39-48-14)19(2)17-36(8,45)33(53-35-29(41)25(40(11)12)16-20(3)49-35)22(5)30(23(6)34(44)51-26)52-27-18-37(9,47-13)32(43)24(7)50-27/h19-27,29-33,35,41-43,45-46H,15-18H2,1-14H3/b39-28+/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OC)\[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyl-tetrahydropyran-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-10-methoxyimino-3,5,7,9,11,13-hexamethyl-oxacyclotetradecan-2-one
Molecular Weight (g/mol) 762.97
Formula Weight 762.4878
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.