missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LFM-A13

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective inhibitor of Bruton's tyrosine kinase (BTK). Inhibits recombinant BTK with an IC50 value of 2.5 μM and has no activity on other protein kinases ( including JAK1, JAK3, HCK, EGFR kinase and insulin receptor kinase) at concentrations of up to 278 μM.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 360 |
| Purity | >98% |
| Molecular Weight (g/mol) | 360.005 |
| Chemical Name or Material | LFM-A13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction